Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Bis(4-methoxycarbonylphenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$809.14 - $809.14

Chemical Identifiers

Chemical Name or Material (R,R)-1,2-Bis(4-methoxycarbonylphenyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C18H20N2O4·2HCl
Formula Weight 401.3
MDL Number MFCD09265307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2596803

  SDS
alfa aesar™
H2596803
1g Each for $809.14
Description & Specifications

Specifications

Formula Weight 401.3
Chemical Name or Material (R,R)-1,2-Bis(4-methoxycarbonylphenyl)-1,2-ethanediamine dihydrochloride
Assay 95%
Assay Percent Notes ee 99%
Molecular Formula C18H20N2O4·2HCl
MDL Number MFCD09265307
Melting Point >200°C
Quantity 1g
Optical Rotation 47° (c=1 Methanol)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.