Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Bis(4-hydroxyphenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$200.41 - $566.54

Chemical Identifiers

Chemical Name or Material (R,R)-1,2-Bis(4-hydroxyphenyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C14H16N2O2·2HCl
Formula Weight 317.23
MDL Number MFCD09265306
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2598203

  SDS
alfa aesar™
H2598203
1g Each for $566.54

AAH25982MD

  SDS
alfa aesar™
H25982MD
250mg Each for $200.41
Description & Specifications

Specifications

Formula Weight 317.23
Health Hazard 1 Warning
Assay 95%
Chemical Name or Material (R,R)-1,2-Bis(4-hydroxyphenyl)-1,2-ethanediamine dihydrochloride
Assay Percent Notes ee 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C14H16N2O2·2HCl
MDL Number MFCD09265306
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.