Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Bis(4-cyanophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$809.14 - $809.14

Chemical Identifiers

Chemical Name or Material (R,R)-1,2-Bis(4-cyanophenyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C16H14N4·2HCl
Formula Weight 335.25
MDL Number MFCD09265296
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2597503

  SDS
alfa aesar™
H2597503
1g Each for $809.14

AAH25975MD

  SDS
alfa aesar™
H25975MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (R,R)-1,2-Bis(4-cyanophenyl)-1,2-ethanediamine dihydrochloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Assay Percent Notes ee 99%
Molecular Formula C16H14N4·2HCl
Formula Weight 335.25
MDL Number MFCD09265296
Melting Point 200°C
Quantity 1g
Optical Rotation 78° (c=1 in Methanol)
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.