missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Bis(4-cyanophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

Manufacturer:  Alfa Aesar™ H25975MD

 View more versions of this product

Catalog No. AAH25975MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(R,R)-1,2-Bis(4-cyanophenyl)-1,2-ethanediamine dihydrochloride
200°C
1055301-12-6
C16H14N4·2HCl
335.25
78° (c=1 in Methanol)
250mg
ee 99%
MFCD09265296
95%

Chemical Identifiers

1055301-12-6
C16H14N4·2HCl
MFCD09265296
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents