missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Bis(2-chlorophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$198.96 - $198.96

Chemical Identifiers

Molecular Formula C14H14Cl2N2·2HCl
Formula Weight 354.12
MDL Number MFCD09265294
Chemical Name or Material (R,R)-1,2-Bis(2-chlorophenyl)-1,2-ethanediamine dihydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH25990MD SDS Alfa Aesar™
H25990MD
250mg Each for $198.96
AAH2599003 SDS Alfa Aesar™
H2599003
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C14H14Cl2N2·2HCl
354.12
MFCD09265294
(R,R)-1,2-Bis(2-chlorophenyl)-1,2-ethanediamine dihydrochloride

Specifications

C14H14Cl2N2·2HCl
MFCD09265294
(R,R)-1,2-Bis(2-chlorophenyl)-1,2-ethanediamine dihydrochloride
354.12
250mg
95%, ee 99%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning