missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-Propylene oxide, ≥98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC399570050

 View more versions of this product

Catalog No. AC399570050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

58.08
0.829g/mL
100.0
C3H6O
98+%
(R)-(+)-Propylene oxide
5g
98.0
(R)-(+)-1, 2-Epoxypropane
MFCD00066211
15448-47-2
Glass bottle

Chemical Identifiers

15448-47-2
100.0
C3H6O
MFCD00066211
(R)-(+)-Propylene oxide
98.0
98+%
58.08
(R)-(+)-1, 2-Epoxypropane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
May cause genetic defects.
Extremely flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger