missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-Propylene Carbonate 98.0+%, TCI America™
$45.34 - $225.16
Chemical Identifiers
CAS | 16606-55-6 |
---|---|
Molecular Formula | C4H6O3 |
Molecular Weight (g/mol) | 102.089 |
MDL Number | MFCD00798265 |
InChI Key | RUOJZAUFBMNUDX-GSVOUGTGSA-N |
Synonym | (R)-4-Methyl-1,3-dioxolan-2-one |
PubChem CID | 641813 |
IUPAC Name | (4R)-4-methyl-1,3-dioxolan-2-one |
SMILES | CC1COC(=O)O1 |
Chemical Identifiers
16606-55-6 | |
102.089 | |
RUOJZAUFBMNUDX-GSVOUGTGSA-N | |
641813 | |
CC1COC(=O)O1 |
C4H6O3 | |
MFCD00798265 | |
(R)-4-Methyl-1,3-dioxolan-2-one | |
(4R)-4-methyl-1,3-dioxolan-2-one |
Specifications
Yellow | |
16606-55-6 | |
MFCD00798265 | |
RUOJZAUFBMNUDX-GSVOUGTGSA-N | |
(4R)-4-methyl-1,3-dioxolan-2-one | |
641813 | |
Clear Liquid at 20°C | |
(R)-Propylene Carbonate |
5g | |
C4H6O3 | |
(R)-4-Methyl-1,3-dioxolan-2-one | |
CC1COC(=O)O1 | |
102.089 | |
102.09 | |
≥98.0% (GC) |
Safety and Handling
EINECSNumber : (5)-0524
RTECSNumber : FF9650000
TSCA : No