missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-Prolinol, 98+%

Manufacturer:  Alfa Aesar™ L1110914

 View more versions of this product

Catalog No. AAL1110914



Specifications

Specifications

(R)-(-)-Prolinol
74°C to 76°C (2mmHg)
86°C (186°F)
C5H11NO
1523669
HVVNJUAVDAZWCB-RXMQYKEDSA-N
[(2R)-pyrrolidin-2-yl]methanol
2724541
101.15
68832-13-3
25g
−31° (c=1 in Toluene)
MFCD00064321
d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, d-prolinol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
C1CC(NC1)CO
101.149
CHEBI:84258
≥98%

Chemical Identifiers

68832-13-3
101.149
HVVNJUAVDAZWCB-RXMQYKEDSA-N
2724541
[(2R)-pyrrolidin-2-yl]methanol
C5H11NO
MFCD00064321
d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, d-prolinol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
CHEBI:84258
C1CC(NC1)CO
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents