missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems ProDots Recombinant Human FGF basic (146 aa)

Pre-aliquoted balls of FGF basic protein for cell culture media

$520.12 - $716.95


Includes: Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human FGF basic.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRD23310 R&D Systems
PRD23310
50μg Each for $520.12
PRD23350 R&D Systems
PRD23350
250μg Each for $716.95
Specifications

Description

ProDots proteins are R&D Systems industry-leading quality, highly bioactive proteins packaged into easy-to-use lyophilized “dots”. Designed to be stable at 4°C and to dissolve instantly, ProDots proteins can be rolled into cell culture media to quickly produce media with a desired working protein concentration.

ProDots proteins are designed to reduce cell culture media preparation time while ensuring optimal protein recovery and bioactivity. Unlike standard lyophilized protein, which is shipped as a diffuse residue within a vial, ProDots proteins are neatly bundled into the shape of a ball. In this form ProDots proteins can be rolled directly into cell culture media. ProDots proteins rehydrate instantly in media to maximize the recovery of purchased protein and to reliably reconstitute it at a desired concentration for use in cell culture media.The accessible storage and high stability of ProDots proteins makes them ideal for quick and easy preparation of cell culture media.

  • Rolls out of the bottle for easy cell culture medium preparation
  • Eliminates time spent aliquoting
  • One ball of FGF basic protein can be used to make 500mL of 10ng/mL cell culture medium
  • Source: E. coli-derived, Pro143-Ser288, with an N-terminal Ala
  • N-terminal Sequence Analysis: Ala-Pro143
  • SDS-PAGE: 17 kDa, reducing conditions
  • Activity: Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749.
  • The ED50 for this effect is typically 0.1-0.6ng/mL.

Order Info

Add ProDots proteins directly to the medium or reconstitute in the vial at 50μg/mL in sterile PBS.