Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-Phanephos, 98%, ee 98%

$211.22 - $211.22

Chemical Identifiers

Chemical Name or Material (R)-Phanephos
CAS 364732-88-7
Molecular Formula C40H34P2
Formula Weight 576.65g/mol
MDL Number MFCD03094575
Synonym (R)-(-)-4,12-Bis(diphenylphosphino)[2.2]paracyclophane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25812MD

 
alfa aesar™
H25812-MD
250mg Each for $211.22
Description & Specifications

Specifications

Melting Point 226° to 230°C
Quantity 250mg
Chemical Name or Material (R)-Phanephos
CAS 364732-88-7
Assay 98%
Assay Percent Notes ee 98%
Molecular Formula C40H34P2
Formula Weight 576.65g/mol
MDL Number MFCD03094575
Synonym (R)-(-)-4,12-Bis(diphenylphosphino)[2.2]paracyclophane