Please login to your online account to display your discounted pricing

(R)-(-)-Niguldipine hydrochloride, Tocris


α1 antagonist, L-type Ca2+ channel blocker; Less active enantiomer of Niguldipine hydrochloride

Quantity 10mg

Manufacturer: r & d systems  RDS112410

 View more versions of this product

Catalog No. 112410

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 646.18
Quantity 10mg
Chemical Name or Material (R)-(-)-Niguldipine hydrochloride
Molecular Formula C36H39N3O6·HCl
CAS 113145-70-3
Assay Percent Range >98%
Synonym (R)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid, 3-(4,4-diphenyl-1-piperidinyl)propyl methyl ester hydrochloride
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material (R)-(-)-Niguldipine hydrochloride
Assay Percent Range >98%
Molecular Formula C36H39N3O6·HCl
Formula Weight 646.18
Synonym (R)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid, 3-(4,4-diphenyl-1-piperidinyl)propyl methyl ester hydrochloride
Accessories

Desiccate at -20°C