Please login to your online account to display your discounted pricing

(R)-(-)-Niguldipine hydrochloride, Tocris

α1 antagonist, L-type Ca2+ channel blocker; Less active enantiomer of Niguldipine hydrochloride

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material (R)-(-)-Niguldipine hydrochloride
Assay Percent Range >98%
Molecular Formula C36H39N3O6·HCl
Formula Weight 646.18
Synonym (R)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid, 3-(4,4-diphenyl-1-piperidinyl)propyl methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

112410

 
r & d systems
RDS112410
10mg Each for $221.01

112450

 
r & d systems
RDS112450
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 646.18
Quantity 10mg
Chemical Name or Material (R)-(-)-Niguldipine hydrochloride
Molecular Formula C36H39N3O6·HCl
CAS 113145-70-3
Assay Percent Range >98%
Synonym (R)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid, 3-(4,4-diphenyl-1-piperidinyl)propyl methyl ester hydrochloride
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C