Please login to your online account to display your discounted pricing

(R)-N-Phthaloyl-2-aminobut-3-en-1-ol, 95%, (95% e.e.), ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-Phthaloyl-2-aminobut-3-en-1-ol
Assay ≥94%
Molecular Formula C12H11NO3
Formula Weight 217.23
MDL Number MFCD06799073
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363182500

  SDS
thermo fisher scientific
AC363182500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 217.23
Quantity 250mg
Color White
Infrared Spectrum Authentic
Enantiomeric Excess 94% min.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (R)-N-Phthaloyl-2-aminobut-3-en-1-ol
Molecular Formula C12H11NO3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD06799073
Packaging Glass bottle
CAS 174810-06-1
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.