Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%

$85.73 - $1,181.72

Chemical Identifiers

Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
CAS 55700-44-2
Molecular Formula C26H28FeNP
Formula Weight 441.34g/mol
MDL Number MFCD00075286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1968504

 
alfa aesar™
L19685-04
2g Each for $1,181.72

AAL19685MC

 
alfa aesar™
L19685-MC
100mg Each for $85.73
Description & Specifications

Specifications

Quantity 2g
Melting Point 141° to 143°C
Quantity 2g
Optical Rotation −355° (c=0.6 in Ethanol)
Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
CAS 55700-44-2
Assay 97%
Molecular Formula C26H28FeNP
Formula Weight 441.34g/mol
MDL Number MFCD00075286
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.