Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate, 95%

Chemical Identifiers

Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate
Molecular Formula C18H25FeNO6
Formula Weight 407.26g/mol
MDL Number MFCD09953453
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2577814

 
alfa aesar™
H25778-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 129° to 131°C
Quantity 25g
Optical Rotation +14° (c=1 in Methanol)
Chemical Name or Material (R)-(-)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate
Assay 95%
Molecular Formula C18H25FeNO6
Formula Weight 407.26g/mol
MDL Number MFCD09953453
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.