Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-N-Methyl-1-phenyl-2-(1-pyrrolidinyl)ethylamine, tech. 90%

Chemical Identifiers

Chemical Name or Material (R)-(-)-N-Methyl-1-phenyl-2-(1-pyrrolidinyl)ethylamine
CAS 136329-39-0
Purity Grade Technical Grade
Molecular Formula C13H20N2
Formula Weight 204.32
MDL Number MFCD01318625
UN Number UN2735
Beilstein 4842022
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1865703

  SDS
alfa aesar™
L1865703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1865706

  SDS
alfa aesar™
L1865706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL18657MD

  SDS
alfa aesar™
L18657MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.32
Chemical Name or Material (R)-(-)-N-Methyl-1-phenyl-2-(1-pyrrolidinyl)ethylamine
CAS 136329-39-0
MDL Number MFCD01318625
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Beilstein 4842022
Molecular Formula C13H20N2
Purity Grade Technical Grade
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Recommended Storage 2° to 8°C
Assay 90%
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Quantity 1g
UN Number UN2735
Optical Rotation −78° (Neat)
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C