missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-N-Isopropyl-1-phenylethylamine hydrochloride, 98+%

$81.58 - $306.88

Chemical Identifiers

CAS 128593-72-6
Molecular Formula C11H18ClN
Molecular Weight (g/mol) 199.722
MDL Number MFCD06011752
InChI Key SVRDLWQPQZOASL-HNCPQSOCSA-N
Synonym r-+-n-isopropyl-1-phenylethylamine hydrochloride, r-n-isopropyl-1-phenylethylamine hydrochloride, isopropyl 1r-1-phenylethyl amine hydrochloride, 1r-1-phenylethyl methylethyl amine, chloride, r-n-2-propyl-1-phenylethylamine hydrochloride, r-n-1-phenylethyl propan-2-amine hydrochloride
PubChem CID 22831491
IUPAC Name N-[(1R)-1-phenylethyl]propan-2-amine;hydrochloride
SMILES CC(C)NC(C)C1=CC=CC=C1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1835406 View Documents Alfa Aesar™
L1835406
5g Each for $306.88
 
AAL1835403 View Documents Alfa Aesar™
L1835403
1g Each for $81.58
 
Specifications

Chemical Identifiers

128593-72-6
199.722
SVRDLWQPQZOASL-HNCPQSOCSA-N
22831491
CC(C)NC(C)C1=CC=CC=C1.Cl
C11H18ClN
MFCD06011752
r-+-n-isopropyl-1-phenylethylamine hydrochloride, r-n-isopropyl-1-phenylethylamine hydrochloride, isopropyl 1r-1-phenylethyl amine hydrochloride, 1r-1-phenylethyl methylethyl amine, chloride, r-n-2-propyl-1-phenylethylamine hydrochloride, r-n-1-phenylethyl propan-2-amine hydrochloride
N-[(1R)-1-phenylethyl]propan-2-amine;hydrochloride

Specifications

262°C to 265°C
128593-72-6
MFCD06011752
r-+-n-isopropyl-1-phenylethylamine hydrochloride, r-n-isopropyl-1-phenylethylamine hydrochloride, isopropyl 1r-1-phenylethyl amine hydrochloride, 1r-1-phenylethyl methylethyl amine, chloride, r-n-2-propyl-1-phenylethylamine hydrochloride, r-n-1-phenylethyl propan-2-amine hydrochloride
CC(C)NC(C)C1=CC=CC=C1.Cl
199.722
199.73
(R)-(+)-N-Isopropyl-1-phenylethylamine hydrochloride
5g
C11H18ClN
3566584
SVRDLWQPQZOASL-HNCPQSOCSA-N
N-[(1R)-1-phenylethyl]propan-2-amine;hydrochloride
22831491
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning