Please login to your online account to display your discounted pricing

(R)-N-FMOC-Styrylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-FMOC-Styrylalanine
Name Note 95%, (98% e.e.)
CAS 215190-23-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C26 H23 N O4
Formula Weight 413.47
MDL Number MFCD01311775
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362555000

  SDS
thermo fisher scientific
362555000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362550010

  SDS
thermo fisher scientific
362550010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 413.47
Quantity 500mg
Color White
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Synonym (R)-N-(9-Fluorenylmethoxycarbonyl)-2-amino-5-phenyl-4-pentenoic acid
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
CAS 215190-23-1
Molecular Formula C26 H23 N O4
Chemical Name or Material (R)-N-FMOC-Styrylalanine
MDL Number MFCD01311775