Learn More
(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 441145000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(R)-N-FMOC-α-Methylvaline | |
616867-28-8,1634-04-4 | |
100.0 | |
Authentic | |
C21H23NO4 | |
-8.6 | |
10% max. (MTBE) | |
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
353.43 | |
353.43 |
98% ee | |
97.5 | |
98.0% min. | |
97.5% min. (HPLC) | |
500mg | |
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid | |
AWEZXIRZNQCCNN-OAQYLSRUSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid | |
11210384 | |
98%,98% ee |
Chemical Identifiers
616867-28-8 | |
353.43 | |
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid |
C21H23NO4 | |
AWEZXIRZNQCCNN-OAQYLSRUSA-N | |
11210384 | |
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only