missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441111000

 View more versions of this product

Catalog No. AC441111000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(R)-N-FMOC-α-Methyl-2-fluorophenylalanine
Authentic
48
111.0°C to 122.0°C
98% ee
97.5
97.5% min. (HPLC)
fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine
CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
419.46
419.46
98%,98% ee
98.0% min.
Glass bottle
White to Yellow
100mg
193086-74-7
100.0
C25H22FNO4
VNYGHKCAXQKZEQ-VWLOTQADSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
44598473
Powder

Chemical Identifiers

193086-74-7
419.46
fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
C25H22FNO4
VNYGHKCAXQKZEQ-VWLOTQADSA-N
44598473
CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Documents