Please login to your online account to display your discounted pricing

(R)-N-FMOC-4-Cyanophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362675000

 View more versions of this product

Catalog No. AC362675000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 412.44
Quantity 500mg
Color Off-white to slightly yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material (R)-N-FMOC-4-Cyanophenylalanine
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C25 H20 N2 O4
MDL Number MFCD00672560
CAS 205526-34-7
Enantiomeric Excess 97.5% min. (HPLC)
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-cyanophenyl-D-alanine
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material (R)-N-FMOC-4-Cyanophenylalanine
Name Note 95%, (98% e.e.)
CAS 205526-34-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C25 H20 N2 O4
Formula Weight 412.44
MDL Number MFCD00672560
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-cyanophenyl-D-alanine
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents