Please login to your online account to display your discounted pricing

(R)-N-FMOC-4-Cyanophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-FMOC-4-Cyanophenylalanine
Name Note 95%, (98% e.e.)
CAS 205526-34-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C25 H20 N2 O4
Formula Weight 412.44
MDL Number MFCD00672560
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362675000

  SDS
thermo fisher scientific
362675000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362670010

  SDS
thermo fisher scientific
362670010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 412.44
Quantity 500mg
Color Off-white to slightly yellow
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Enantiomeric Excess 97.5% min. (HPLC)
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-cyanophenyl-D-alanine
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Molecular Formula C25 H20 N2 O4
MDL Number MFCD00672560
Name Note 95%, (98% e.e.)
Chemical Name or Material (R)-N-FMOC-4-Cyanophenylalanine
CAS 205526-34-7
Assay Percent Range 95%
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.