Please login to your online account to display your discounted pricing

(R)-N-FMOC-4-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362470010

 View more versions of this product

Catalog No. AC362470010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 466.32
Quantity 1g
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-bromophenyl-D-alanine
Chemical Name or Material (R)-N-FMOC-4-Bromophenylalanine
Assay Percent Range 95%
CAS 198545-76-5
MDL Number MFCD00273459
Name Note 95%, (98% e.e.)
Molecular Formula C24 H20 Br N O4

Specifications

Chemical Name or Material (R)-N-FMOC-4-Bromophenylalanine
Name Note 95%, (98% e.e.)
CAS 198545-76-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
Formula Weight 466.32
MDL Number MFCD00273459
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-bromophenyl-D-alanine