Please login to your online account to display your discounted pricing

(R)-N-FMOC-4-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-FMOC-4-Bromophenylalanine
Name Note 95%, (98% e.e.)
CAS 198545-76-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C24 H20 Br N O4
Formula Weight 466.32
MDL Number MFCD00273459
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362475000

  SDS
thermo fisher scientific
362475000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362470010

  SDS
thermo fisher scientific
362470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 466.32
Physical Form Powder
Quantity 500mg
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-bromophenyl-D-alanine
Enantiomeric Excess 97.5% min. (HPLC)
Packaging Glass bottle
Chemical Name or Material (R)-N-FMOC-4-Bromophenylalanine
Infrared Spectrum Authentic
Assay Percent Range 95%
MDL Number MFCD00273459
Molecular Formula C24 H20 Br N O4
CAS 198545-76-5
Name Note 95%, (98% e.e.)