Please login to your online account to display your discounted pricing

(R)-N-FMOC-3-Thienylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-FMOC-3-Thienylalanine
Name Note 95%, (98% e.e.)
CAS 220497-90-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C22 H19 N O4 S
Formula Weight 393.46
MDL Number MFCD00151915
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362575000

  SDS
thermo fisher scientific
362575000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 393.46
Physical Form Crystalline Powder
Quantity 500mg
Color Off-white
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Synonym N-(9-Fluorenylmethoxycarbonyl)-3-thienyl-D-alanine
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C22 H19 N O4 S
Name Note 95%, (98% e.e.)
MDL Number MFCD00151915
CAS 220497-90-5
Chemical Name or Material (R)-N-FMOC-3-Thienylalanine