Please login to your online account to display your discounted pricing

(R)-N-FMOC-(2-Furyl)alanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362490010

 View more versions of this product

Catalog No. AC362490010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 377.39
Quantity 1g
Color White to light yellow
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Enantiomeric Excess 97.5% min. (HPLC)
Synonym N-(9-Fluorenylmethoxycarbonyl)-2-furyl-D-alanine
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material (R)-N-FMOC-(2-Furyl)alanine
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
Molecular Formula C22H19NO5
MDL Number MFCD01311774
CAS 220497-85-8

Specifications

Chemical Name or Material (R)-N-FMOC-(2-Furyl)alanine
Name Note 95%, (98% e.e.)
CAS 220497-85-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C22H19NO5
Formula Weight 377.39
MDL Number MFCD01311774
Synonym N-(9-Fluorenylmethoxycarbonyl)-2-furyl-D-alanine