Please login to your online account to display your discounted pricing

(R)-N-FMOC-2-(5-Bromothienyl)alanine, 95%, 98% e.e., ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362485000

 View more versions of this product

Catalog No. AC362485000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 472.35
Quantity 500mg
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym N-(9-Fluorenylmethoxycarbonyl)-5-bromo-2-thienyl-D-alanine
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material (R)-N-FMOC-2-(5-Bromothienyl)alanine
Assay Percent Range 95%
Name Note 95%, (98% e.e.)
Molecular Formula C22H18BrNO4S
CAS 220497-83-6
MDL Number MFCD01311772

Specifications

Chemical Name or Material (R)-N-FMOC-2-(5-Bromothienyl)alanine
Name Note 95%, (98% e.e.)
CAS 220497-83-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C22H18BrNO4S
Formula Weight 472.35
MDL Number MFCD01311772
Synonym N-(9-Fluorenylmethoxycarbonyl)-5-bromo-2-thienyl-D-alanine