Please login to your online account to display your discounted pricing

(R)-N-FMOC-2-(5-Bromothienyl)alanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-FMOC-2-(5-Bromothienyl)alanine
Name Note 95%, (98% e.e.)
CAS 220497-83-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C22H18BrNO4S
Formula Weight 472.35
MDL Number MFCD01311772
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362480010

  SDS
thermo fisher scientific
362480010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362485000

  SDS
thermo fisher scientific
362485000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 472.35
Quantity 1g
Color White to off-white
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Synonym N-(9-Fluorenylmethoxycarbonyl)-5-bromo-2-thienyl-D-alanine
MDL Number MFCD01311772
Chemical Name or Material (R)-N-FMOC-2-(5-Bromothienyl)alanine
Name Note 95%, (98% e.e.)
CAS 220497-83-6
Molecular Formula C22H18BrNO4S
Assay Percent Range 95%