Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-N-Diphenylphosphino-N-methyl-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, (R)-Methyl BoPhoz™

Asymmetric hydrogenation

$69.72 - $69.72

Chemical Identifiers

Chemical Name or Material (R)-N-Diphenylphosphino-N-methyl-[ (S)-2- (diphenylphosphino)ferrocenyl]ethylamine, (R)-Methyl BoPhoz™
CAS 406680-94-2
Molecular Formula C37H35FeNP2
Formula Weight 611.50g/mol
MDL Number MFCD05865220
Synonym (R)-Methyl BoPhoz™
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4468477

 
alfa aesar™
44684-77
0.1g Each for $69.72
Description & Specifications

Specifications

Color Yellow to orange
Melting Point 112° to 115°C
Quantity 0.1g
Odor Odorless
Optical Rotation −297° (C=1 in dichloromethane)
Chemical Name or Material (R)-N-Diphenylphosphino-N-methyl-[ (S)-2- (diphenylphosphino)ferrocenyl]ethylamine, (R)-Methyl BoPhoz™
CAS 406680-94-2
Physical Form Solid
Molecular Formula C37H35FeNP2
Formula Weight 611.50g/mol
MDL Number MFCD05865220
Synonym (R)-Methyl BoPhoz™