missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-N,α-Dimethylbenzylamine, ≥99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC395850050

 View more versions of this product

Catalog No. AC395850050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(R)-(+)-N α-Dimethylbenzylamine
Authentic
5g
99.0
Glass bottle
+63° to +65° (20°C, 589nm) (C=4, methanol)
≥99.0%
C9H13N
135.21
0.9240g/mL
Clear colorless
5933-40-4
100.0
1.5100 to 1.5120 (20°C, 589nm)
Liquid
99+%
C6H5CH(CH3)NHCH3
MFCD00044966

Chemical Identifiers

5933-40-4
100.0
C9H13N
135.21
(R)-(+)-N α-Dimethylbenzylamine
99.0
99+%
C6H5CH(CH3)NHCH3
MFCD00044966
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger