Please login to your online account to display your discounted pricing

(R)-(+)-N,α-Dimethylbenzylamine, ≥99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC395850010

 View more versions of this product

Catalog No. AC395850010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 135.21
Assay ≥99.0%
Quantity 1g
Color Clear colorless
Density 0.9240g/mL
CAS Min % 99.0
CAS Max % 100.0
Linear Formula C6H5CH(CH3)NHCH3
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C9H13N
Assay Percent Range 99+%
MDL Number MFCD00044966
Chemical Name or Material (R)-(+)-N α-Dimethylbenzylamine
CAS 5933-40-4
Specific Rotation +63° to +65° (20°C, 589nm) (C=4, methanol)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

Specifications

Chemical Name or Material (R)-(+)-N α-Dimethylbenzylamine
CAS 5933-40-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H13N
Linear Formula C6H5CH(CH3)NHCH3
Formula Weight 135.21
MDL Number MFCD00044966
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

SDS & Safety Documents