Please login to your online account to display your discounted pricing

(R)-N-BOC-Propargylglycine, 95%, 98% e.e., ACROS Organics™

$646.83 - $646.83

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-Propargylglycine
Name Note 98% ee
CAS 63039-46-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H15NO4
Formula Weight 213.23
MDL Number MFCD01320886
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362850050

  SDS
thermo fisher scientific
AC362850050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362850010

  SDS
thermo fisher scientific
362850010
Glass bottle 1g Each for $646.83
Description & Specifications

Specifications

Formula Weight 213.23
Assay ≥94%
Physical Form Crystalline Powder
Quantity 5g
Color Off-white to light beige
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material (R)-N-BOC-Propargylglycine
Assay Percent Range 95%
CAS 63039-46-3
Molecular Formula C10H15NO4
Name Note 98% ee
MDL Number MFCD01320886
Enantiomeric Excess 97.5% min. (GC)
Packaging Glass bottle