missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-N-BOC-piperidine-3-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 163438-09-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
InChI Key NXILIHONWRXHFA-MRVPVSSYSA-N
Synonym boc-d-nip-oh, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-1-boc-nipecotic acid, r-1-boc-piperidine-3-carboxylic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-boc-nip, r-boc-nip-oh, r-boc-nipecotic acid, r-n-boc-piperidine-3-carboxylic acid
PubChem CID 643516
IUPAC Name (3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436500010
SDS View Documents
Thermo Fisher Scientific
AC436500010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436500050
SDS View Documents
Thermo Fisher Scientific
AC436500050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

163438-09-3
229.28
boc-d-nip-oh, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-1-boc-nipecotic acid, r-1-boc-piperidine-3-carboxylic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-boc-nip, r-boc-nip-oh, r-boc-nipecotic acid, r-n-boc-piperidine-3-carboxylic acid
(3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
C11H19NO4
NXILIHONWRXHFA-MRVPVSSYSA-N
643516
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O

Specifications

Authentic
−52.20° (20°C c=1.15 MeOH)
White
1g
96.0
96% min. (HPLC)
boc-d-nip-oh, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-1-boc-nipecotic acid, r-1-boc-piperidine-3-carboxylic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-boc-nip, r-boc-nip-oh, r-boc-nipecotic acid, r-n-boc-piperidine-3-carboxylic acid
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
229.28
229.28
97%
Glass bottle
-52.2
166°C to 171°C
163438-09-3
100.0
C11H19NO4
NXILIHONWRXHFA-MRVPVSSYSA-N
(3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
643516
Crystalline Powder
(R)-N-BOC-Piperidine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS