Please login to your online account to display your discounted pricing

(R)-N-BOC-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362910010

 View more versions of this product

Catalog No. AC362910010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 196707-32-1
CAS Min % 94.0
CAS Max % 100.0
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
TSCA TSCA
Packaging Glass bottle
Physical Form Powder
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H16FNO4
Formula Weight 269.27
MDL Number MFCD01320889
Quantity 1g
Color White
Synonym (D)-N-tert-Butoxycarbonyl-α-(4-fluorophenyl)acetic acid

Specifications

Chemical Name or Material (R)-N-BOC-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 196707-32-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H16FNO4
Formula Weight 269.27
MDL Number MFCD01320889
Synonym (D)-N-tert-Butoxycarbonyl-α-(4-fluorophenyl)acetic acid