Please login to your online account to display your discounted pricing

(R)-N-BOC-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-4-Fluorophenylglycine
Name Note 95%, (98% e.e.)
CAS 196707-32-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H16FNO4
Formula Weight 269.27
MDL Number MFCD01320889
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362910010

  SDS
thermo fisher scientific
362910010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 269.27
Quantity 1g
Color White
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
MDL Number MFCD01320889
Name Note 95%, (98% e.e.)
Molecular Formula C13H16FNO4
Assay Percent Range 95%
Chemical Name or Material (R)-N-BOC-4-Fluorophenylglycine
CAS 196707-32-1
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym (D)-N-tert-Butoxycarbonyl-α-(4-fluorophenyl)acetic acid