Please login to your online account to display your discounted pricing

(R)-N-BOC-4-Fluorophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  364430010

 View more versions of this product

Catalog No. AC364430010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Quantity 1g
Color White
TSCA TSCA
Packaging Glass bottle
CAS 57292-45-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Enantiomeric Excess 97.5% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (R)-N-BOC-4-Fluorophenylalanine
Molecular Formula C14 H18 F N O4
Formula Weight 283.30
MDL Number MFCD00079669
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym N-tert-Butoxycarbonyl-4-fluorophenyl-D-alanine

Specifications

Chemical Name or Material (R)-N-BOC-4-Fluorophenylalanine
Name Note 95%, (98% e.e.)
CAS 57292-45-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 F N O4
Formula Weight 283.30
MDL Number MFCD00079669
Synonym N-tert-Butoxycarbonyl-4-fluorophenyl-D-alanine
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents