Please login to your online account to display your discounted pricing

(R)-N-BOC-4-Fluorophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-4-Fluorophenylalanine
Name Note 95%, (98% e.e.)
CAS 57292-45-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 F N O4
Formula Weight 283.30
MDL Number MFCD00079669
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC364430010

  SDS
thermo fisher scientific
364430010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Quantity 1g
Color White
CAS 57292-45-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Glass bottle
Enantiomeric Excess 97.5% min.
Chemical Name or Material (R)-N-BOC-4-Fluorophenylalanine
Formula Weight 283.30
Molecular Formula C14 H18 F N O4
MDL Number MFCD00079669
Assay Percent Range 95%
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Name Note 95%, (98% e.e.)
Synonym N-tert-Butoxycarbonyl-4-fluorophenyl-D-alanine
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.