Please login to your online account to display your discounted pricing

(R)-N-BOC-4-Chlorophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  362920050

 View more versions of this product

Catalog No. AC362920050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 299.75
Quantity 5g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (R)-N-BOC-4-Chlorophenylalanine
Name Note 95%, (98% e.e.)
MDL Number MFCD00076978
CAS 57292-44-1
Assay Percent Range 95%
Molecular Formula C14H18ClNO4
Packaging Glass bottle
Synonym N-tert-Butoxycarbonyl-4-chlorophenyl-D-alanine
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (R)-N-BOC-4-Chlorophenylalanine
Name Note 95%, (98% e.e.)
CAS 57292-44-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H18ClNO4
Formula Weight 299.75
MDL Number MFCD00076978
Synonym N-tert-Butoxycarbonyl-4-chlorophenyl-D-alanine