Please login to your online account to display your discounted pricing

(R)-(-)-N-BOC-3-pyrrolidinol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396860050

 View more versions of this product

Catalog No. AC396860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.24
Physical Form Powder
Assay ≥97.5%
Quantity 5g
Color White to off-white
CAS Max % 100.0
CAS 109431-87-0
Synonym (R)-(-)-N-(tert-Butoxycarbonyl)-3-pyrrolidinol
Chemical Name or Material (R)-(-)-N-BOC-3-pyrrolidinol
Assay Percent Range 98%
Molecular Formula C9H17NO3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01317838
CAS Min % 97.5
Packaging Glass bottle
Specific Rotation -25° to -27° (20°C, 589nm) (C=1, methanol)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material (R)-(-)-N-BOC-3-pyrrolidinol
CAS 109431-87-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H17NO3
Formula Weight 187.24
MDL Number MFCD01317838
Synonym (R)-(-)-N-(tert-Butoxycarbonyl)-3-pyrrolidinol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents