missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-N-BOC-3-pyrrolidinol, 98%, ACROS Organics™

Chemical Identifiers

CAS 109431-87-0
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD01317838
InChI Key APCBTRDHCDOPNY-SSDOTTSWSA-N
Synonym r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol
PubChem CID 6544479
IUPAC Name tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC396860010 SDS View Documents Acros Organics
AC396860010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC396860050 SDS View Documents Acros Organics
AC396860050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

109431-87-0
187.239
APCBTRDHCDOPNY-SSDOTTSWSA-N
6544479
CC(C)(C)OC(=O)N1CCC(C1)O
C9H17NO3
MFCD01317838
r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol
tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate

Specifications

Authentic
-25° to -27° (20°C, 589nm) (C=1, methanol)
1g
97.5
98%
MFCD01317838
APCBTRDHCDOPNY-SSDOTTSWSA-N
tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate
6544479
Powder
(R)-(-)-N-BOC-3-pyrrolidinol
Glass bottle
White
109431-87-0
100.0
C9H17NO3
r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol
CC(C)(C)OC(=O)N1CCC(C1)O
187.239
187.24
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.