Please login to your online account to display your discounted pricing

(R)-N-BOC-3-Fluorophenylalanine, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-3-Fluorophenylalanine
Name Note 98% ee
CAS 114873-11-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 F N O4
Formula Weight 283.3
MDL Number MFCD00672523
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362880010

  SDS
thermo fisher scientific
362880010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 283.3
Quantity 1g
Color Off-white
Packaging Glass bottle
CAS 114873-11-9
Chemical Name or Material (R)-N-BOC-3-Fluorophenylalanine
Name Note 98% ee
Molecular Formula C14 H18 F N O4
Assay Percent Range 95%
Enantiomeric Excess 97.5% min.
MDL Number MFCD00672523
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic