Please login to your online account to display your discounted pricing

(R)-N-BOC-3-Fluorophenylalanine, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362880010

 View more versions of this product

Catalog No. AC362880010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 283.3
Quantity 1g
Color Off-white
Chemical Name or Material (R)-N-BOC-3-Fluorophenylalanine
Name Note 98% ee
CAS 114873-11-9
Packaging Glass bottle
Enantiomeric Excess 97.5% min.
Assay Percent Range 95%
Infrared Spectrum Authentic
Molecular Formula C14 H18 F N O4
MDL Number MFCD00672523
CAS Min % 94.0
CAS Max % 100.0

Specifications

Chemical Name or Material (R)-N-BOC-3-Fluorophenylalanine
Name Note 98% ee
CAS 114873-11-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14 H18 F N O4
Formula Weight 283.3
MDL Number MFCD00672523