Please login to your online account to display your discounted pricing

(R)-N-BOC-3-Amino-3-phenylpropanoic acid, 95%, 98% e.e., ACROS Organics™

$506.01 - $506.01

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-3-Amino-3-phenylpropanoic acid
Name Note 98% ee
CAS 161024-80-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H19NO4
Formula Weight 265.31
MDL Number MFCD01320859
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362862500

  SDS
thermo fisher scientific
AC362862500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362860010

  SDS
thermo fisher scientific
362860010
Glass bottle 1g Each for $506.01

AC362865000

  SDS
thermo fisher scientific
362865000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 265.31
Quantity 250mg
Color White
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (R)-N-BOC-3-Amino-3-phenylpropanoic acid
CAS 161024-80-2
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Molecular Formula C14H19NO4
Name Note 98% ee
Assay Percent Range 95%
MDL Number MFCD01320859
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.