Please login to your online account to display your discounted pricing

(R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  362870010

 View more versions of this product

Catalog No. AC362870010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 344.2
Physical Form Powder
Quantity 1g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym (R)-N-BOC-β-(4-bromophenyl)-β-alanine
Chemical Name or Material (R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
Molecular Formula C14H18BrNO4
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
MDL Number MFCD01320860
CAS 261380-20-5
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min.

Specifications

Chemical Name or Material (R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
CAS 261380-20-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H18BrNO4
Formula Weight 344.2
MDL Number MFCD01320860
Synonym (R)-N-BOC-β-(4-bromophenyl)-β-alanine