Please login to your online account to display your discounted pricing

(R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
Name Note 95%, (98% e.e.)
CAS 261380-20-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H18BrNO4
Formula Weight 344.2
MDL Number MFCD01320860
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362870010

  SDS
thermo fisher scientific
362870010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362872500

  SDS
thermo fisher scientific
362872500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 344.2
Physical Form Powder
Quantity 1g
Color White
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Synonym (R)-N-BOC-β-(4-bromophenyl)-β-alanine
Chemical Name or Material (R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
Molecular Formula C14H18BrNO4
Name Note 95%, (98% e.e.)
Assay Percent Range 95%
MDL Number MFCD01320860
CAS 261380-20-5
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min.