Please login to your online account to display your discounted pricing

(R)-(+)-N-BOC-2-Piperidinecarboxylic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399250010

 View more versions of this product

Catalog No. AC399250010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5% (Acid)
Formula Weight 229.27
Quantity 1g
Color White to brown
Melting Point 120° to 125°C
CAS Max % 100.0
Chemical Name or Material (R)-(+)-N-BOC-2-Piperidinecarboxylic acid
Assay Percent Range 98%
Specific Rotation +62.0° to +65.0° (20°C, 589nm) (c=1%, acetic acid)
Molecular Formula C11H19NO4
CAS 28697-17-8
MDL Number MFCD00237380
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (R)-(+)-N-BOC-2-Piperidinecarboxylic acid
CAS 28697-17-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (Acid)
Assay Percent Range 98%
Molecular Formula C11H19NO4
Formula Weight 229.27
MDL Number MFCD00237380
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents