Please login to your online account to display your discounted pricing

(R)-N-BOC-2-(5-Bromothienyl)alanine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material (R)-N-BOC-2-(5-Bromothienyl)alanine
Assay Percent Range 95%
Molecular Formula C12H16BrNO4S
Formula Weight 350.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC362775000

  SDS
thermo fisher scientific
AC362775000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC362770010

  SDS
thermo fisher scientific
AC362770010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (R)-N-BOC-2-(5-Bromothienyl)alanine
CAS 261380-16-9
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C12H16BrNO4S
Formula Weight 350.23
Quantity 500mg