Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-N-Benzyl-1-phenylethylamine, ChiPros 99+%, ee 96+%

$52.43 - $158.22

Chemical Identifiers

Chemical Name or Material (R)-(+)-N-Benzyl-1-phenylethylamine
CAS 38235-77-7
Molecular Formula C15H17N
Formula Weight 211.31
MDL Number MFCD00015010
Beilstein 3589990
Synonym D-(+)-N-Benzyl-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1929806

  SDS
alfa aesar™
L1929806
5g Each for $52.43

AAL1929814

  SDS
alfa aesar™
L1929814
25g Each for $158.22
Description & Specifications

Specifications

Formula Weight 211.31
Chemical Name or Material (R)-(+)-N-Benzyl-1-phenylethylamine
CAS 38235-77-7
Assay ≥99%
Beilstein 3589990
Optical Rotation +39° (Neat)
Flash Point >110°C (230°F)
Molecular Formula C15H17N
Quantity 5g
MDL Number MFCD00015010
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym D-(+)-N-Benzyl-alpha-methylbenzylamine
Assay Percent Notes ee ≥96%
Boiling Point 171°C (15mmHg)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.