missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(+)-N-(alpha-Methylbenzyl)phthalamic Acid, TCI America™
$117.64 - $397.23
Chemical Identifiers
CAS | 21752-35-2 |
---|---|
Molecular Formula | C16H15NO3 |
Molecular Weight (g/mol) | 269.3 |
MDL Number | MFCD00063050 |
InChI Key | VCFKXWGKKDZMPO-LLVKDONJSA-N |
Synonym | r-2-1-phenylethyl carbamoyl benzoic acid, r-+-n-1-phenylethyl phthalamic acid, 2-1r-1-phenylethyl carbamoyl benzoic acid, r-+-n-alpha-methylbenzyl phthalamic acid, r-o-1-phenylethyl amino carbonyl benzoic acid, benzoic acid,2-1r-1-phenylethyl amino carbonyl, benzoic acid, 2-1r-1-phenylethyl amino carbonyl, 2-1r-1-phenylethyl carbamoyl benzoic acid, +-n-1-phenylethyl phthalic acid mon, r-+-alpha-methylbenzyl phthalamic acid |
PubChem CID | 89038 |
IUPAC Name | 2-[[(1R)-1-phenylethyl]carbamoyl]benzoic acid |
SMILES | CC(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2C(=O)O |
Chemical Identifiers
21752-35-2 | |
269.3 | |
VCFKXWGKKDZMPO-LLVKDONJSA-N | |
89038 | |
CC(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2C(=O)O |
C16H15NO3 | |
MFCD00063050 | |
r-2-1-phenylethyl carbamoyl benzoic acid, r-+-n-1-phenylethyl phthalamic acid, 2-1r-1-phenylethyl carbamoyl benzoic acid, r-+-n-alpha-methylbenzyl phthalamic acid, r-o-1-phenylethyl amino carbonyl benzoic acid, benzoic acid,2-1r-1-phenylethyl amino carbonyl, benzoic acid, 2-1r-1-phenylethyl amino carbonyl, 2-1r-1-phenylethyl carbamoyl benzoic acid, +-n-1-phenylethyl phthalic acid mon, r-+-alpha-methylbenzyl phthalamic acid | |
2-[[(1R)-1-phenylethyl]carbamoyl]benzoic acid |
Specifications
White | |
1g | |
C16H15NO3 | |
r-2-1-phenylethyl carbamoyl benzoic acid, r-+-n-1-phenylethyl phthalamic acid, 2-1r-1-phenylethyl carbamoyl benzoic acid, r-+-n-alpha-methylbenzyl phthalamic acid, r-o-1-phenylethyl amino carbonyl benzoic acid, benzoic acid,2-1r-1-phenylethyl amino carbonyl, benzoic acid, 2-1r-1-phenylethyl amino carbonyl, 2-1r-1-phenylethyl carbamoyl benzoic acid, +-n-1-phenylethyl phthalic acid mon, r-+-alpha-methylbenzyl phthalamic acid | |
CC(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2C(=O)O | |
269.3 | |
269.30 | |
≥98.0% (HPLC,T) |
128°C | |
21752-35-2 | |
MFCD00063050 | |
VCFKXWGKKDZMPO-LLVKDONJSA-N | |
2-[[(1R)-1-phenylethyl]carbamoyl]benzoic acid | |
89038 | |
Crystal-Powder at 20°C | |
(R)-(+)-N-(alpha-Methylbenzyl)phthalamic Acid |
Safety and Handling
TSCA : No