Learn More
(R)-(+)-N alpha-Dimethylbenzylamine, 99+%, Thermo Scientific™
Chemical Identifiers
CAS | 5933-40-4 |
---|---|
Molecular Formula | C9H13N |
Molecular Weight (g/mol) | 135.21 |
MDL Number | MFCD00044966 |
InChI Key | RCSSHZGQHHEHPZ-MRVPVSSYSA-N |
Synonym | 1r-n-methyl-1-phenylethanamine, a-dimethylbenzylamine, benzenemethanamine, n,alpha-dimethyl-, alphar, methyl 1r-1-phenylethyl amine, r-+-n,, r-+-n,a-dimethylbenzylamine, r-+-n,alpha-dimethylbenzylamine, r-+-n-methyl-1-phenylethylamine, r-n-methyl-1-phenylethanamine, r-n-methyl-alpha-phenylethylamine, unii-7iz3431sc3 |
PubChem CID | 2060888 |
IUPAC Name | (1R)-N-methyl-1-phenylethanamine |
SMILES | CC(C1=CC=CC=C1)NC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC395850010
|
Thermo Fisher Scientific
AC395850010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC395850050
|
Thermo Fisher Scientific
AC395850050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5933-40-4 | |
135.21 | |
RCSSHZGQHHEHPZ-MRVPVSSYSA-N | |
2060888 | |
CC(C1=CC=CC=C1)NC |
C9H13N | |
MFCD00044966 | |
1r-n-methyl-1-phenylethanamine, a-dimethylbenzylamine, benzenemethanamine, n,alpha-dimethyl-, alphar, methyl 1r-1-phenylethyl amine, r-+-n,, r-+-n,a-dimethylbenzylamine, r-+-n,alpha-dimethylbenzylamine, r-+-n-methyl-1-phenylethylamine, r-n-methyl-1-phenylethanamine, r-n-methyl-alpha-phenylethylamine, unii-7iz3431sc3 | |
(1R)-N-methyl-1-phenylethanamine |
Specifications
5933-40-4 | |
100.0 | |
Colorless | |
61°C | |
Authentic | |
C6H5CH(CH3)NHCH3 | |
Glass bottle | |
1r-n-methyl-1-phenylethanamine, a-dimethylbenzylamine, benzenemethanamine, n,alpha-dimethyl-, alphar, methyl 1r-1-phenylethyl amine, r-+-n,, r-+-n,a-dimethylbenzylamine, r-+-n,alpha-dimethylbenzylamine, r-+-n-methyl-1-phenylethylamine, r-n-methyl-1-phenylethanamine, r-n-methyl-alpha-phenylethylamine, unii-7iz3431sc3 | |
Solubility in water: insoluble | |
0.924 | |
(1R)-N-methyl-1-phenylethanamine | |
135.21 | |
135.21 | |
99+% |
99.0 | |
0.9240g/mL | |
74.0°C to 76.0°C (11.0mmHg) | |
99% min. (GC) | |
C9H13N | |
MFCD00044966 | |
1.5100 to 1.5120 (20°C, 589nm) | |
1g | |
RCSSHZGQHHEHPZ-MRVPVSSYSA-N | |
CC(C1=CC=CC=C1)NC | |
+63° to +65° (20°C, 589nm) (C=4, methanol) | |
2060888 | |
Liquid | |
(R)-(+)-N α-Dimethylbenzylamine |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
RUO – Research Use Only